Learn More
Thermo Scientific™ Sterilin™ Autoclave Bags
Sterilin Autoclave Bags are strong disposable bags with blue biohazard printing specifically designed for contaminated waste disposal in autoclaves or incinerators.
Brand: Thermo Scientific™ 511-17
329.77 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : 56
Additional Details : Weight : 5.90400kg
Description
Sterilin Autoclave Bags are strong disposable bags with blue biohazard printing specifically designed for contaminated waste disposal in autoclaves or incinerators.
- HDPE bags are suitable for sterilization at 121°C and polypropylene bags are suitable for the inactivation of particularly resistant biological waste at 135°C
- Convenient “tissue box” cartons dispense bags individually
- Coated wire stands or cardboard holders available for use with autoclave bags
Ordering Alerts:
Not available in all geographic regions
Specifications
Autoclave bags (HDPE) | |
200 | |
810 x 610mm | |
810 mm | |
610 mm |
HDPE | |
121°C | |
32 in. | |
24 in. |