missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sterilin™ BenchGuard
GREENER_CHOICE

Protect benches and other surfaces against liquid spills with the highly absorbent Thermo Scientific™ Sterilin™ BenchGuard.

Brand:  Thermo Scientific™ BG60E

134.68 GBP valid until 2024-12-31
Use promo code "21745" to get your promotional price.


 View more versions of this product

Product Code. 11399183

  • £265.00 / 50 sheets
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

Protect benches and other surfaces against liquid spills with the highly absorbent Thermo Scientific™ Sterilin™ BenchGuard. Absorbent liner also provides a cushion for glassware when accidentally dropped on benchtop.

  • Available in both 400mL/m2 and 800mL/m2absorption capacities
  • Plastic coated on one side to prevent soak through
  • Available in roll or sheet form
  • Rolls supplied in easy to use dispenser packs
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Bench Guard
1.607612 ft.
1.6076 ft.
BenchGuard Extra
800 mL/m2
0.6 m
0.49 m
50 x Sheets
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.