missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sterilin™ Clear Microtiter™ Plates

Store and transport samples or perform microbiology screenings, EIA/absorbance assays and serology work with 96-well Thermo Scientific™ Sterilin™ Clear Microtiter™ Plates.

Brand:  Thermo Scientific™ 611F96

150.91 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.



Code : 56

Additional Details : Weight : 2.39400kg

 View more versions of this product

Product Code. 11349163

  • £192.00 / Pack of 50
In Stock
to see stock.
Add to basket

Description

Description

Store and transport samples or perform microbiology screenings, EIA/absorbance assays and serology work with 96-well Thermo Scientific™ Sterilin™ Clear Microtiter™ Plates. Supplied without lid; may be purchased separately. Not available in all geographic regions.

  • Manufactured from virgin polystyrene, giving excellent optical clarity
  • Aseptically manufactured plates receive no surface treatment and therefore provide a medium binding surface
  • Irradiation process for the sterile plates makes the surface more hydrophilic, therefore generating a high binding product
  • Suitable for use at a wavelength of 340nm
  • Condensation rings on the lid, together with raised well rims on the base, help minimize the risk of contamination from surrounding wells
  • Orientation corners and alphanumeric labeling ensure easy sample identification
  • Frosted write-on area on end wall of plate for clear identification
  • Hanging well design ensures even temperature distribution around each well
  • The plates are supplied without a lid (can be purchased separately)
  • In vitro use only

Ordering Alerts:

Not available in all geographic regions

Specifications

Specifications

Standard
8 x 12
Clear
Polystyrene
Non-sterile
Flat Bottom (F-well)
5
Microtiter Plates, Clear, Flat bottom, NS
50 Cases
Microplate
96
Non-treated
50
50/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers