Learn More
Thermo Scientific™ Sterilin™ Clear Microtiter™ Plates
Store and transport samples or perform microbiology screenings, EIA/absorbance assays and serology work with 96-well Thermo Scientific™ Sterilin™ Clear Microtiter™ Plates.
£130.00 - £278.00
Specifications
Color | Clear |
---|---|
Product Type | Microplate |
Material | Polystyrene |
No. of Wells | 96 |
No. per Case | 50 |
Product Code | Brand | Description | Sterility | No. per Case | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Sterility | No. per Case | Price | Quantity & Availability | |||
11339163
|
Thermo Scientific™
611U96 |
Microtiter Plates, Clear, U bottom, NS | Non-sterile | 50 |
£187.00
Pack of 50 |
In Stock
Log in to see stock. |
|||
11897403
|
Thermo Scientific™
611V96 |
Microtiter Plates, Clear, V bottom | Non-sterile | 50 |
£176.00
Pack of 50 |
In Stock
Log in to see stock. |
|||
11349163
|
Thermo Scientific™
611F96 |
Microtiter Plates, Clear, Flat bottom, NS | Non-sterile | 50 |
£199.00
Pack of 50 |
In Stock
Log in to see stock. |
|||
11399153
|
Thermo Scientific™
612U96 |
Microtiter Plates, Clear, U bottom | Sterile | 50 |
£234.00
Pack of 50 |
In Stock
Log in to see stock. |
|||
11309163
|
Thermo Scientific™
612V96 |
Microtiter Plates, Clear, V bottom | Sterile | 50 |
£278.00
Pack of 50 |
Estimated Shipment: 04-02-2025
Log in to see stock. |
|||
11319163
|
Thermo Scientific™
612F96 |
Microtiter Plates, Clear, Flat bottom | Sterile | 50 |
£130.00
Pack of 50 |
In Stock
Log in to see stock. |
|||
11329163
|
Thermo Scientific™
642000 |
Plate Lid | Sterile | 50 |
£132.00
Pack of 50 |
In Stock
Log in to see stock. |
|||
Description
Store and transport samples or perform microbiology screenings, EIA/absorbance assays and serology work with 96-well Thermo Scientific™ Sterilin™ Clear Microtiter™ Plates. Supplied without lid; may be purchased separately. Not available in all geographic regions.
Features
- Manufactured from virgin polystyrene for excellent optical clarity
- Manufactured in ISO Class 7 Clean Room without surface treatment for a medium binding surface
- Available irradiated for sterility and a more hydrophilic surface, providing high binding
- Suitable for use at a wavelength of 340nm
- Minimizes the risk of contamination from surrounding wells with condensation rings on the lid raised well rims on the base.
- Ensures even temperature distribution around each well with a hanging well design
Easy Sample Identification
- Frosted writing area on the end wall
- Orientation corners
- Alphanumeric labeling
Note:
Plates are supplied without a lid (can be purchased separately).
Ordering Information:
Product is not available in all geographic regions.
Specifications
Clear | |
Polystyrene | |
50 |
Microplate | |
96 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.