Learn More
Thermo Scientific™ Sterilin™ Loops and Spreaders
Simplify your culture work with Thermo Scientific™ Sterilin™ Loops and Spreaders. Choices for inoculation, dilution streaking, spreading and colony picking.
£76.77 - £669.75
Product Code | Brand | Product Type | Volume | Color | Quantity | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Volume | Color | Quantity | Price | Quantity & Availability | ||
11379283
|
Thermo Scientific™
SL1H |
Inoculating Loop and Needle, Rigid | 1 μL | Dark green | 10 | N/A | |||
11834732
|
Thermo Scientific™
SL1S |
Inoculating Loop and Needle, Flexible | 1 μL | Pale Green | 10 | N/A | |||
11369283
|
Thermo Scientific™
SL10H |
Inoculating Loop and Needle, Rigid | 10 μL | Dark blue | 10 | N/A | |||
11814732
|
Thermo Scientific™
SL10S |
Inoculating Loop and Needle, Flexible | 10 μL | Pale Blue | 10 | N/A | |||
11778858
|
Thermo Scientific™
QL1 |
Inoculating Loop, Spherical Tip | 1 μL | Green | 1000 | N/A | |||
11788858
|
Thermo Scientific™
QL10 |
Inoculating Loop, Spherical Tip | 10 μL | Blue | 1000 | N/A | |||
11718868
|
Thermo Scientific™
SN20 |
Inoculating Needle | Violet | 1000 | N/A | ||||
11788978
|
Thermo Scientific™
SPCS01 |
L-Shaped Spreader | Blue | 500 | N/A | ||||
11728868
|
Thermo Scientific™
SPCS05 |
L-Shaped Spreader | Blue | 1000 | N/A | ||||
Description
Simplify your culture work with Thermo Scientific™ Sterilin™ Loops and Spreaders. Choices for inoculation, dilution streaking, spreading and colony picking.
NOT AVAILABLE IN AMERICA
- For inoculation, dilution streaking, spreading and picking isolated colonies
- Eliminate the need for flaming and the subsequent risk of aerosols Inoculation loop for fixed sample volumes with choice of sphere for dilution streaking, or needle for picking colonies and stab cultures
- Choice of rigid or flexible loops to suit different applications and user preferences
- Hexagonal shaft improves grip and assists orientation
- Free of lubricants, oils and electrostatic charges to facilitate consistent wetting and complete liquid transfer
- No rough edges means smooth, problem-free plating and streaking of cultures
- Sterilized by gamma irradiation
- Not available in all geographic regions
Specifications
Irradiated |