missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sterilin™ Plain Swabs

Product Code. 11329633
Click to view available options
For Use With (Application):
General Purpose
Applicator Material:
Wood
Packaging:
Peel pouch
No. per Case:
2500
Unit Size:
Pack of 2500
This item is not returnable. View return policy

Product Code. 11329633

Brand: Thermo Scientific™ 165KS100

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Simplify sample collection with Thermo Scientific™ Sterilin™ Plain Swabs. Ideal for samples that will be processed immediately and don't require preservative for transport.

Simplify sample collection with Thermo Scientific™ Sterilin™ Plain Swabs. Ideal for samples that will be processed immediately and don't require preservative for transport. Supplied in either a peel pouch or in a tube for ease of handling and transportation.

AVAILABLE IN UK ONLY

  • Primary sample collection device for bacteria
  • Color-coded caps for ease of identification

Certifications

  • CE marked as Class IIa in accordance with the Medical Device Directive 93/42/EEC (for transient invasive use)

Notes

  • Must be processed as soon as possible after sampling (ideally within two-hour period) to avoid dehydration of the sample and possible nonrecovery of pathogens

Ordering Information

  • Not available in all geographic regions

Specifica

Sterility EO Sterile
Applicator Material Wood
Certifications/Compliance CE Marked: Class IIa, Medical Device Directive 93/42/EEC
Tip Material Cotton
Packaging Peel pouch
For Use With (Application) General Purpose
No. per Pack 100
No. per Case 2500
Cap Color N/A
Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato