Learn More
Thermo Scientific™ Sterilin™ Plastic Round Base Tubes
Avoid the hazards of glass with this safe, convenient alternative. Thermo Scientific™ Sterilin™ Plastic Round Base Tubes are manufactured from virgin polystyrene.
Brand: Thermo Scientific™ RT15
515.00 GBP valid until 2024-12-31
Use promo code "23291" to get your promotional price.
Code : 56
Additional Details : Weight : 6.40000kg
Description
Avoid the hazards of glass with this safe, convenient alternative. Thermo Scientific™ Sterilin™ Plastic Round Base Tubes are manufactured from virgin polystyrene and supplied nonsterile. Not available in all geographic regions.
- Suitable for samples ranging from 0.6 to 11mL
- Supplied nonsterile
Specifications
Base Tube | |
Polystyrene | |
1000 | |
Non-sterile |
Round Base Plastic Tubes | |
6 mm | |
Sterilin™ | |
0.6 mL |