missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sterilin™ Plastic Round Base Tubes

Avoid the hazards of glass with this safe, convenient alternative. Thermo Scientific™ Sterilin™ Plastic Round Base Tubes are manufactured from virgin polystyrene.

Brand:  Thermo Scientific™ RT20

512.00 GBP valid until 2024-12-31
Use promo code "23291" to get your promotional price.



Code : 56

Additional Details : Weight : 7.54000kg

 View more versions of this product

Product Code. 13015723

  • £655.00 / Pack of 10000
Estimated Shipment: 21-10-2024
to see stock.
Add to basket

Description

Description

Avoid the hazards of glass with this safe, convenient alternative. Thermo Scientific™ Sterilin™ Plastic Round Base Tubes are manufactured from virgin polystyrene and supplied nonsterile. Not available in all geographic regions.

  • Suitable for samples ranging from 0.6 to 11mL
  • Supplied nonsterile
Specifications

Specifications

Round Base Plastic Tubes
6 mm
Non-sterile
Polystyrene
1000
1.0 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers