Learn More
Thermo Scientific™ Sterilin™ Polystyrene Containers, 60mL to 250mL
Contain samples in Thermo Scientific™ Sterilin™ Polystyrene Containers with 60mL to 250mL capacities. Available with either plastic or metal flow seal caps.
£165.00 - £672.00
Product Code | Brand | Capacity (Metric) | Closure Material | Label | No. per Case | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Closure Material | Label | No. per Case | Price | Quantity & Availability | ||
11379123
|
Thermo Scientific™
125AM |
60 mL | Metal | No Label | 300/Cs. |
£524.00
Pack of 300 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11319143
|
Thermo Scientific™
125BM |
60 mL | Metal | Printed | 300/Cs. |
£519.00
Pack of 300 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11339153
|
Thermo Scientific™
125CM |
60 mL | Metal | Plain | 300/Cs. |
£606.00
Pack of 300 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11389123
|
Thermo Scientific™
125AP |
60 mL | Polyethylene | No Label | 300/Cs. |
£165.00
Pack of 300 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11788898
|
Thermo Scientific™
125BP |
60 mL | Polyethylene | Printed | 300/Cs. |
£565.00
Pack of 300 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11708908
|
Thermo Scientific™
125CP |
60 mL | Polyethylene | Plain | 300/Cs. |
£550.00
Pack of 300 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11399123
|
Thermo Scientific™
185AM |
100 mL | Metal | No Label | 200/Cs. |
£570.00
Pack of 200 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11349143
|
Thermo Scientific™
185BM |
100 mL | Metal | Printed | 200/Cs. |
£247.68
Pack of 200 |
Estimated Shipment: 02-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11369143
|
Thermo Scientific™
185CM |
100 mL | Metal | Plain | 200/Cs. |
£672.00
Pack of 200 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11309133
|
Thermo Scientific™
185AP |
100 mL | Polyethylene | No Label | 200/Cs. |
£590.00
Pack of 200 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11379143
|
Thermo Scientific™
185CP |
100 mL | Polyethylene | Plain | 200/Cs. |
£662.00
Pack of 200 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11319133
|
Thermo Scientific™
165A |
150 mL | Metal | No Label | 120/Cs. |
£431.00
Pack of 120 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11389143
|
Thermo Scientific™
165B |
150 mL | Metal | Printed | 120/Cs. |
£443.00
Pack of 120 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11399143
|
Thermo Scientific™
165C |
150 mL | Metal | Plain | 120/Cs. |
£430.00
Pack of 120 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11329133
|
Thermo Scientific™
190A |
250 mL | Metal | No Label | 50/Cs. |
£214.66
Pack of 50 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11349153
|
Thermo Scientific™
190B |
250 mL | Metal | Printed | 50/Cs. |
£250.00
Pack of 50 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
12047492
|
Thermo Scientific™
190C |
250 mL | Metal | Plain | 50/Cs. |
£236.00
Pack of 50 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11359143
|
Thermo Scientific™
185BP |
100 mL | Polyethylene | Printed | 200/Cs. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
Contain samples in Thermo Scientific™ Sterilin™ Polystyrene Containers with 60mL to 250mL capacities. Available with either plastic or metal flow seal caps.
NOT AVAILABLE IN AMERICA
- Leak-tested in accordance with BS EN ISO 6717
- 125AM, 125BM and 125CM product certified 95kPa-compliant ensuring suitability for sample transport
Ordering Information:
Product is not available in North America.
Specifications
Container | |
Yes | |
Polystyrene |