Learn More
Thermo Scientific™ Sterilin™ Test Tube Holders
Protect your test tubes and their contents with our flexible-cavity Thermo Scientific™ Sterilin™ Test Tube Holders. Unique design prevents them from floating in water baths.
Brand: Thermo Scientific™ BLKTTS5
Description
Protect your test tubes and their contents with our flexible-cavity Thermo Scientific™ Sterilin™ Test Tube Holders. Unique design prevents them from floating in water baths.
- Unique rubber design with flexible cavity diameters
- Will sink in water baths and grips tubes to prevent them from floating to the surface
- Not available in all geographic regions
Specifications
Tube Rack | |
Rubber | |
180 × 105mm | |
28.5 mm |
Tubes (28.5 mm) | |
15 | |
28.5 mm |
Note: To ensure a secure grip, please select a cavity that is 1.5-2.0mm smaller than the tube diameter (e.g., 12.5mm cavity holds a 14mm diameter tube securely)