missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sterilin™ UTM (Universal Transport Medium) Swabs
GREENER_CHOICE

Simplify collection, transport, maintenance and frozen storage of virus, chlamydia, mycoplasma and ureaplasma specimens. Available as filled tube with separate swab or in a kit.

Specifications

Applicator Material Plastic
Certifications/Compliance CE Marked as Class IIa in accordance with the Medical Device Directive93/42/EEC (for transient invasive use)
No. per Case 50
View More Specs

Products 9
Product Code Brand Description Sterility No. per Case Price Quantity & Availability  
Product Code Brand Description Sterility No. per Case Price Quantity & Availability  
12842843
View Documents
Thermo Scientific™
305C
Collection kit, 3mL medium, nasopharyngeal flocked swab Irradiated 50 N/A
N/A
12892843
View Documents
Thermo Scientific™
328C
Collection kit, 3mL medium + regular dacron swab Aseptically Manufactured/Gamma-irradiated 50 N/A
N/A
12852843
View Documents
Thermo Scientific™
330C
Bulk tubes, 3mL medium Aseptically Manufactured 50 N/A
N/A
12706588
View Documents
Thermo Scientific™
346C
Collection kit, 3mL medium + regular flocked swab Aseptically Manufactured/Gamma-irradiated 50 N/A
N/A
13157673
View Documents
Thermo Scientific™
347C
Collection kit, 3mL medium, neonatal flocked swab Aseptically Manufactured/Gamma-irradiated 50 N/A
N/A
13187673
View Documents
Thermo Scientific™
350C
Bulk tubes, 1mL medium Aseptically Manufactured 50 N/A
N/A
12874641
View Documents
Thermo Scientific™
359C
1mL medium, regular flocked swab Aseptically Manufactured/Gamma-irradiated 50 N/A
N/A
13137683
GREENER_CHOICE
View Documents
Thermo Scientific™
360C
1mL medium, nasopharyngeal flocked swab Irradiated 50 N/A
N/A
13147683
View Documents
Thermo Scientific™
361C
1mL medium, minitip flocked swab Irradiated 50 N/A
N/A
Description

Description

Simplify collection, transport, maintenance and frozen storage of virus, chlamydia, mycoplasma and ureaplasma specimens. Suitable for cell culture, Rapid Antigen Detection, DFA, EIA, PCR and nucleic acid amplification assays. Thermo Scientific™ Sterilin™ UTM (Universal Transport Medium) Swabs are available as filled tube with separate swab or in a kit.
AVAILABLE IN UK ONLY

  • Room temperature stable and incorporates a cryoprotectant
  • Formulation includes antibiotics to inhibit bacterial and fungal flora in patient samples
  • Free-standing polypropylene skirted tube with conical base enabling convenient sample preparation—simply vortex and inoculate direct into tissue culture
  • Available as filled tubes with a choice of separate individually wrapped swabs or a collection kit with the swab included
  • Glass beads in the tube help with the release and dispersion of the patient sample from the swab
  • All swabs feature molded breakpoint to enable safe, reliable breaking of the swab into the tube—enables the regular swab to be captured by the cap as it is replaced on the tube
Certifications
  • CE marked as Class IIa in accordance with the Medical Device Directive 93/42/EEC (for transient invasive use)
Ordering Information
  • Available in UK only
Specifications

Specifications

Plastic
CE Marked as Class IIa in accordance with the Medical Device Directive93/42/EEC (for transient invasive use)
50
Videos
SDS
Documents

Documents

Product Certifications