missing translation for 'onlineSavingsMsg'
Learn More

Brand™ Self-adhesive Sterilization Indicator Tape

Contains thermo-active pigments. BRAND™ Self-adhesive Sterilization Indicator Tape turns brown after autoclaving.

Brand:  Brand™ 61750

12.34 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.


Product Code. 10480863

  • £19.25 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

Self-adhesive crêpe paper containing thermo-active pigments. The white colored strips turn brown after autoclaving (121°C: after approximately 20 min, or 134°C: after approximately 5 min.)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

50 m x 19 mm
50 m
121°C to 134°C
Yes
19 mm
Detecting sterilization completion
Tape Strips
Crepe paper with thermo-active pigment
121° 20 min., 134° 5 min.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24155

    Expires2025-04-30

    Hot Deal

    *Discount will apply to qualifying orders received by Fisher Scientific between 01/03/2025 and 31/03/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.

    Terms & Conditions See Full Promo Details
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Brand™ Self-adhesive Sterilization Indicator Tape > Sterilization temperature: 121 to 134deg.C; Dimensions (LxW): 50m x 19mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.