Learn More
DWK Life Sciences STERIPLAN™ Petri Dish, from Soda-lime Glass
Petri dishes are used for the cultivation of bacteria, yeasts, plant and animal cells on solid media.
£24.85 - £174.00
Specifications
Autoclavable | Not Autoclavable |
---|---|
Material | Soda Lime Glass |
For Use With (Application) | Growth and cultivation of microorganisms; Cultivation on solid gel culture medium; Microscopy of microorganisms; Cell culture |
No. per Pack | 10/Pk. |
Product Code | Brand | Diameter (Metric) | Height (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Height (Metric) | Price | Quantity & Availability | ||||
11740834
|
DWK Life Sciences
237554008 |
60 mm | 15 mm |
£30.55
Pack of 10 |
In Stock
Log in to see stock. |
||||
11760834
|
DWK Life Sciences
237554505 |
100 mm | 10 mm |
£33.65
Pack of 10 |
Estimated Shipment: 13-06-2024
Log in to see stock. |
||||
11750844
|
DWK Life Sciences
237554608 |
100 mm | 15 mm |
£34.62
Pack of 10 |
In Stock
Log in to see stock. |
||||
11760844
|
DWK Life Sciences
237554805 |
100 mm | 20 mm |
£34.62
Pack of 10 |
Estimated Shipment: 18-06-2024
Log in to see stock. |
||||
11770844
|
DWK Life Sciences
237555107 |
120 mm | 20 mm |
£52.80
Pack of 10 |
In Stock
Log in to see stock. |
||||
11780844
|
DWK Life Sciences
237555201 |
150 mm | 25 mm |
£85.24
Pack of 10 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
11735424
|
DWK Life Sciences
237555904 |
200 mm | 30 mm |
£118.00
Pack of 10 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
11756968
|
DWK Life Sciences
237553903 |
40 mm | 12 mm |
£30.80
Pack of 10 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
11766968
|
DWK Life Sciences
237555604 |
180 mm | 30 mm |
£174.00
Pack of 10 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
11750834
|
DWK Life Sciences
237554205 |
80 mm | 15 mm |
£30.75
Pack of 10 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
11688683
|
DWK Life Sciences
1184071 |
90 mm | 15 mm |
£24.85
Pack of 10 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
11776968
|
DWK Life Sciences
237556103 |
200 mm | 45 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Cultivation of microorganisms can take place on a gel-like nutrient medium, which is placed in a flat, round, transparent dish with lid (the so-called Petri dish). Due to the solid gel, mixing of the microorganisms is prevented and larger cell culture accumulations (colonies) can grow.
- Free from bubbles and streaks
- Even distribution of sewing bottom possible
Specifications
Not Autoclavable | |
Soda Lime Glass | |
Growth and cultivation of microorganisms; Cultivation on solid gel culture medium; Microscopy of microorganisms; Cell culture | |
10/Pk. |