missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ SteriPlast™ Blue Food Scoop
GREENER_CHOICEBIOPHARMA

Can be used to take samples of powders, granulates and pastes. The long handle makes it possible to sample larger containers even with little content or to reach through narrow openings.

£29.65 - £31.50

Specifications

Product Type Shovel
Material Polystyrene
Autoclavable Not Autoclavable
Handle Type Long-handled
Disposable Single-use
View More Specs

Products 2
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
17260604
GREENER_CHOICEBIOPHARMA
View Documents
Buerkle™
5378-5015
50 mL
£29.65
Pack of 10
In Stock
to see stock.
Add to basket
 
17200614
GREENER_CHOICEBIOPHARMA
View Documents
Buerkle™
5378-5019
100 mL
£31.50
Pack of 10
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
 
Description

Description

  • With the straight bottom of the scoop, the medium can be scooped up directly from the bottom of the container
  • PS, white
  • Individually packaged and optionally sterilised
  • Handle length 300 mm
  • For single use
  • Clean room manufactured (clean room class 7)
  • Complies with EU food requirements and FDA regulations
Specifications

Specifications

Shovel
Not Autoclavable
Single-use
Blue
10
Polystyrene
Long-handled
Sterile
Complies with EU food rrequirements and FDA regulations
300 mm
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.