missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buerkle™ SteriPlast™ Blue Detectable Spoons
For sampling powders, granulates and fluids. Buerkle™ SteriPlast™ Blue Detectable Spoons are sterilized by gamma rays.
£57.45 - £63.70
Specifications
Product Type | Detectable Sampling Spoon |
---|---|
Material | Polystyrene |
Autoclavable | Not Autoclavable |
Disposable | Yes |
Sterility | Sterile |
Product Code | Brand | Length (Metric) | Capacity (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Capacity (Metric) | Price | Quantity & Availability | ||||
15684114
|
Buerkle™
5378-2011 |
127 mm | 2.5 mL |
£57.45
Pack of 100 |
In Stock
Log in to see stock. |
||||
15694114
|
Buerkle™
5378-2012 |
170 mm | 10 mL |
£63.70
Pack of 100 |
In Stock
Log in to see stock. |
||||
Description
- Use of blue tools helps to meet requirements of HACCP hazard analysis and assessment in reference to foreign objects
- This is important criterion in food processing and production, and in animal feed and pharmaceutical industry
- Can be used as part of HACCP/IFS foreign object management
- Individually packaged for disposable use
- Production, assembly and packaging according to clean room class 7
- Can be detected by colour scanners, metal detectors or X-rays
Specifications
Detectable Sampling Spoon | |
Not Autoclavable | |
Sterile | |
Sampling Powders, Granulates and Fluids |
Polystyrene | |
Yes | |
Blue |