missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ SteriPlast™ DispoLadle
GREENER_CHOICEBIOPHARMA

Embossed for measuring in 10mL increments. Buerkle™ SteriPlast™ DispoLadle is specifically designed for disposable use. Ergonomic handle on both sides for right or left-hand operation.

Brand:  Buerkle™ 5393-6651

181.79 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 15604144

  • £205.00 / Pack of 20
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • 200mL dipper
  • 92mm external diameter
  • Ergonomic handle
  • Spouts on both sides for right or left-hand operation
  • HDPE
  • Clean room manufactured, individually packaged
  • Optionally sterilized by gamma rays
  • 20 pieces per pack
Specifications

Specifications

Ladle
200 mL
Sterile
White
HDPE
Single-use
Dual spouts
1000 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.