missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ SteriPlast™ Blue Food Scoops
GREENER_CHOICEBIOPHARMA

Product Code. p-7083204 Shop All Bürkle Products
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
150 mL
250 mL
500 mL
1000 mL
2000 mL
Length (Metric):
141 mm
160 mm
205 mm
216 mm
232 mm
280 mm
332 mm
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 15614134

Brand: Buerkle™ 53783013

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

For sampling powders, granulates and pastes. Buerkle™ SteriPlast™ Food Scoops are sterilized by gamma rays. The color blue does not occur in a natural form in foodstuffs. Scoops can be quickly found and easily identified in a visual check if ever mixed into production.

  • Use of blue tools helps to meet requirements of HACCP hazard analysis and assessment in reference to foreign objects
  • This is important criterion in food processing and production, and in animal feed and pharmaceutical industry
  • Can be used as part of HACCP/IFS foreign object management
  • Individually packaged for disposable use
  • Production, assembly and packaging according to clean room class 7
  • Can be detected by colour scanners

Compliance

EU food requirements

TRUSTED_SUSTAINABILITY

Specifications

Sterility Sterile
Autoclavable Not Autoclavable
Capacity (English) 8.45 oz.
Capacity (Metric) 250 mL
Color Blue
Disposable Single-use
Product Type Food Sampling Scoop
For Use With (Application) Sampling Powders, Granulates and Pastes
Length (English) 9.1 in.
Material Polystyrene
Length (Metric) 232 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.