Learn More
Buerkle™ SteriPlast™ Micro Spatulas
GREENER_CHOICEBIOPHARMA
Product Code. 15624114 Shop All Bürkle Products
Click to view available options
Capacity (Metric):
1 mL, 0.25 mL
Unit Size:
Pack of 100
Product Code. 15624114
Brand: Buerkle™ 53781032
Only null left
Add to basket
Edge
Add to basket
Use to sample, transfer or process small amounts of chemicals, powders, granulates, pastes, creams or liquids. Buerkle™ SterilPlast™ Micro Spatulas facilitates precise work with sturdy centre handle.
  • Polystyrene
  • Production, assembly and packaging according to clean room class 7
  • Complies with EU food requirements and FDA regulations
  • Packaged individually and sterilized by gamma rays
TRUSTED_SUSTAINABILITY
Specifications
Sterility Sterile
Capacity (Metric) 1 mL, 0.25 mL
Certifications/Compliance EU food regulations
Material Polystyrene