missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ SteriPlast™ Disposable Sample Scoop
GREENER_CHOICEBIOPHARMA

Sample powder, granules, and pastes with these sterile polystyrene scoops. Available with or without transparent lids to help ensure a sterile and contamination-free sampling procedure.

Brand:  Buerkle™ 5378-1020

 View more versions of this product

Product Code. 17280584

  • £86.85 / Pack of 10
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Available with or without a transparent cover to help protect samples from contamination
  • Ergonomically designed handle with a slightly angled edge
  • Scoop stands horizontally and can be laid down without materials trickling out
  • Ideal fo ruse with powders, granules, and pastes
  • Individually packed for one-way needs
  • Sterilized by gamma radiation
  • Production, assembling, and packaging according to cleanroom class 7 (10.000)
Specifications

Specifications

Disposable Scoop
Not Autoclavable
2000 mL
Sterile
Polystyrene
67.63 oz.
Single-use
Complies with EU food rrequirements and FDA regulations
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number244277 Certificate TypeDeclaration of Conformity Date19/11/2024 Catalog Number5378-1020
Lot Number241391 Certificate TypeCertificate of Compliance Date29/04/2024 Catalog Number5378-1020
Lot Number234630 Certificate TypeCertificate of Compliance Date13/11/2023 Catalog Number5378-1020
Lot Number233163 Certificate TypeCertificate of Compliance Date25/07/2023 Catalog Number5378-1020
Lot Number224871 Certificate TypeCertificate of Compliance Date13/12/2022 Catalog Number5378-1020
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Buerkle™ SteriPlast™ Disposable Sample Scoop > 2000 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.