Learn More
Buerkle™ SteriPlast™ Blue Spatula
For sampling powders, granulates and pastes. Buerkle™ SteriPlast™ Spatula for Foodstuffs is sterilized by gamma rays.
Brand: Buerkle™ 5378-3009
17.08 GBP valid until 2024-12-31
Use promo code "23267" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.17000kg
Description
- Long, ergonomically-shaped, stable handle and sharp blade
- Penetrate directly into containers such as paper or plastic sacks
- Use of blue tools helps to meet requirements of HACCP hazard analysis and assessment in reference to foreign objects
- This is important criterion in food processing and production, and in animal feed and pharmaceutical industry
- Can be used as part of HACCP/IFS foreign object management
- Individually packaged for disposable use
- Production, assembly and packaging according to clean room class 7
- Can be detected by colour scanners
Compliance
EU food requirements
Specifications
Food Spatula | |
Long, ergonomically-shaped, stable handle | |
Sterile | |
Sampling Powders, Granulates and Pastes | |
263 mm |
Polystyrene | |
Yes | |
Blue | |
10.3 in. |