man
Test
Search
Order Status
Quick Order
Sign In
chevron-down
chevron-down
Learn More
Home
Products
Sterilizers and Autoclaves
Sterilization Verification Indicators
12908015
Print
Steris™ Spordex™ Biological Indicators
Geobacillus Stearothermophilus E6; D-val: 1.5 to 2.5; Qty: 100 count
Brand:
Steris™
Additional Details :
Weight :
12.00000kg
Product Code.
12908015
£1125.00
/ Pack of 100
Quantity
Estimated Shipment: 06-08-2024
Log in