missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Stedim Biotech™ Sterisart Bd Bottled Media: TSB, Infusion Bottle
Sterility testing fluid TSB is used to detect, isolate, and cultivate fastidious and non-fastidious microorganisms
Brand: Sartorius Stedim Biotech™ BD-257247
Additional Details : Weight : 3.20000kg
Description
- Tryptic soy broth
- A general purpose liquid nutrient medium used in sterility testing for the detection, isolation, and cultivation of fastidious and non fastidious microorganisms acc. to USP and EP.
- Nutritious medium that will support the growth of a wide variety of microorganisms, including common aerobic and facultatively anaerobic bacteria and fungi.
Specifications
Type 1 Closure, 32 mm OD | |
Sterile | |
125 mL Bottle |
TSB (Tryptic Soy Broth) | |
25 | |
100 mL |
Safety and Handling
Product Identifier
- X25 Sterisart BD bottled Media, Type: TSB