missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Stedim Biotech™ Sterisart Bd Bottled Media: TSB, Wide-Mouth Bottle

Sterility testing fluid TSB is used to detect, isolate, and cultivate fastidious and non-fastidious microorganisms

Brand:  Sartorius Stedim Biotech™ BD-257294

Additional Details : Weight : 3.20000kg

Product Code. 15982710

  • £262.00 / Pack of 10
Estimated Shipment: 06-09-2024
to see stock.
Add to basket

Description

Description

  • Tryptic soy broth
  • A general purpose liquid nutrient medium used in sterility testing for the detection, isolation, and cultivation of fastidious and non fastidious microorganisms acc. to USP and EP.
  • Nutritious medium that will support the growth of a wide variety of microorganisms, including common aerobic and facultatively anaerobic bacteria and fungi.
Specifications

Specifications

Type 7 Closure, 45 mm OD
Sterile
Wide-mouth bottle, individually double-packaged
TSB (Tryptic Soy Broth)
10
100 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers