Learn More
Sartorius Sterisart NF gamma, Transfersystem
The Sterisart™ NF System is a completely disposable technology based on the membrane filtration method, for the sterility testing of pharmaceutical products.
Brand:  Sartorius 16472--------GBD
Product Code. 17827960
  • £329.00 / Pack of 10
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Unique Septum Version for aseptic sampling
  • High retention of microbes
  • Low adsorption
  • High mechanical stability
  • Sharp Metal needle for easy penetration
  • Pre-installed color-coded tube clamps
  • Easy-to-read graduated marks
  • User-friendly (several practical adapters available)
  • Product-/lot number identification
  • Huge variety of adapters and more than 20 different unit types
  • Gas-impermeable packaging for protection against sterilants
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Dual Needle
Sterile
Tubing system with a dual needle
Glass
Sterility Testing