Learn More
Sampling Systems™ SteriWare™ Cupped V Powder Spatulas Type 2
Improve your quality control by taking fixed volume powder samples easily with Type 2 SteriWare™ Cupped V Powder Spatulas. The unique cupped end allows you to accurately weigh samples. Available sterile in a variety of lengths and widths. Ideal for pharmaceutical, food and chemical industries.
£99.90 - £139.00
Specifications
Product Type | Powder Spatula |
---|---|
Material | Polystyrene |
Color | White |
Quantity | 100 Pk. |
Product Code | Brand | Length (Metric) Blade | Width (Metric) Blade | Size | Sterility | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) Blade | Width (Metric) Blade | Size | Sterility | Price | Quantity & Availability | ||
17534606
|
Sampling Systems™
8115A-100 |
100 mm | 20 mm | Standard | Non-sterile |
£103.00
Pack of 100 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||
17544606
|
Sampling Systems™
8115A-100S |
100 mm | 20 mm | Standard | Sterile |
£123.00
Pack of 100 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||
17554606
|
Sampling Systems™
8115A-150S |
150 mm | 20 mm | Standard | Sterile |
£125.00
Pack of 100 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||
17564606
|
Sampling Systems™
8115A-250 |
250 mm | 20 mm | Standard | Non-sterile |
£110.00
Pack of 100 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||
17574606
|
Sampling Systems™
8115A-250S |
250 mm | 20 mm | Standard | Sterile |
£139.00
Pack of 100 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||
17584606
|
Sampling Systems™
8115N-100 |
100 mm | 10 mm | Narrow | Non-sterile |
£99.90
Pack of 100 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||
17594606
|
Sampling Systems™
8115N-100S |
100 mm | 10 mm | Narrow | Sterile |
£120.00
Pack of 100 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||
17504616
|
Sampling Systems™
8115N-150 |
150 mm | 10 mm | Narrow | Non-sterile |
£103.00
Pack of 100 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||
17514616
|
Sampling Systems™
8115N-150S |
150 mm | 10 mm | Narrow | Sterile |
£122.00
Pack of 100 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||
17524616
|
Sampling Systems™
8115N-250 |
250 mm | 10 mm | Narrow | Non-sterile |
£104.00
Pack of 100 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||
17534616
|
Sampling Systems™
8115N-250S |
250 mm | 10 mm | Narrow | Sterile |
£134.00
Pack of 100 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||
Description
- The disposable V spatula is ideal for powder and harder material sampling.
- It is commonly used for surface sampling, raw material testing and general goods in testing.
- The cupped design means it samples a fixed volume.
- It also stops powder flowing out which frequently happens with traditional open ended spatula designs.
- Manufactured to the highest standard in a medical cleanroom to ensure cleanliness and supplied with full documentation.
- Available in 2 widths and 3 lengths to ensure you have the right sampling tool for the job.
- The narrow versions are ideal if you want to put the sample into the test tube or take a small sample.
- The longer version is ideal when you want to take more of a cross sectional sample.
- The disposable V spatula comes pre bagged and ready to use.
- Simply unwrap and use.
- No cleaning or cleaning validation is required.
- Available in 2 versions, either sterile or non sterile versions.
- It comes with an ergonomic handle and it has a free standing design.
- Sterile and non sterile spatulas are ideal when a robust tool is required.
- Made from high quality PS that conforms to the latest food grade, EC and FDA standards.
- BSE/TSE free.
- Batch certificates of conformity supplied.
- Moulded and Packed in a medical cleanroom to maintain cleanliness.
- Simply unwrap and use.
Specifications
Powder Spatula | |
Polystyrene | |
White | |
100 Pk. |