Learn More
Sampling Systems™ SteriWare™ Disposable Graduated Jugs
![BIOPHARMA](/content/dam/fishersci/glyphs/sure_trace.png)
Click to view available options
Quantity:
35 Non-Sterile
35 Sterile
Description
- Moulded and packed in a Class 100,000 Cleanroom
- Individually bagged
- Made from FDA, EC 1935/2004 and EU 10/2011 conforming material
- BSE/TSE free
- Graduation marks in mL
- Full batch traceability
- Available pre-sterilized (SAL 10-6)
Specifications
Capacity | 500mL |
Material | Polypropylene |
Quantity | 35 Non-Sterile |
Sterility | Non-sterile |
Graduated | Yes |