missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ SteriWare™ Disposable HDPE Powder Lances

Sampling Systems™ SteriWare™ Disposable HDPE Powder Lances are ideal for taking a cross sectional sample through a bulk powder.

Brand:  Sampling Systems™ 8063H-300

Code : Z4

Additional Details : Weight : 2.00000kg

Product Code. 12947945

  • £81.60 / Pack of 20
Estimated Shipment: 19-08-2024
to see stock.
Add to basket

Description

Description

  • Assembled and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming HDPE
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • Suitable for use in ATEX areas
  • Push the sampler into the product, rotate the sampler then withdraw
  • Deposit the sample into a suitable container
  • Place the used sampler into its original plastic bag and dispose
  • No cross contaminatin
  • No cleaning required
Specifications

Specifications

HDPE
75 mL
400 mm
20 per box
Non-sterile
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers