Learn More
Sampling Systems™ SteriWare™ Disposable Liquid Funnel
Ideal for laboratories, pharmaceutical environments, cleanrooms and chemical manufactruing. Sampling Systems™ SteriWare™ Disposable Liquid Funnel comes individually bagged after being moulded and packed in a Class 100,000 Cleanroom.
Brand: Sampling Systems™ 8370H-100
This item is not returnable.
View return policy
Description
- Constructed of HDPE
- Moulded and packed in a Class 100,000 Cleanroom
- Individually bagged
- Made from FDA, EC 1935/2004 and EU 10/2011 conforming material
- BSE/TSE free
- Full batch traceability
- Available pre-sterilized (SAL 10-6)
- No cross contamination
- No cleaning required
Specifications
100 mm | |
12 mm | |
Non-sterile | |
Liquid Funnel |