missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ SteriWare™ Disposable Sack Sampler

Product Code. 15691582 Shop All Sampling Systems Products
Click to view available options
Width (Metric):
14 mm
25 mm
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 15691582

Brand: Sampling Systems™ 8085N100S

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Sampling Systems™ SteriWare™ Disposable Sack Sampler is a hygienic, single use device that has been specially designed to take samples through the side of the sack.

  • Individually bagged
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming materials
  • BSE/TSE free
  • Full batch traceability
  • Suitable for use in ATEX areas
  • Available pre-sterilized (SAL 10-6)

Specifications

Material HDPE
Sterility Sterile
Length (Metric) 400 mm
Width (Metric) 14 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.