Learn More
Sampling Systems™ SteriWare™ Forceps (Tweezers)
Shop All Sampling Systems Products
Click to view available options
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 100
Description
- Our disposable forceps are available in both sterile and non sterile versions.
- They are suitable for lifting and moving small objects such as tablets, samples etc.
- The handles have undulations moulded into them to give you extra grip.
- These lightweight tweezers are suitable for food, chemical, pharmaceutical, or cosmetic applications.
- These tweezers can be used straight from the bag.
- The sterile tweezers have been processed by gamma irradiation.
- Made from food grade PS.
- Conforms to the latest FDA and EU requirements.
- BSE/TSE free
- To avoid contamination, they have been manufactured and then packed in a class 7 cleanroom.
- They come with full documentation that confirms food grade and sterility.
Specifications
Specifications
| Material | Polystyrene |
| Length (Metric) | 115 mm |
| Sterility | Non-sterile |
| Packaging Type | Bag |