Learn More
Sampling Systems™ SteriWare™ Forceps (Tweezers)
Our Sterile single use SteriWare™ Forceps (Tweezers) are completely manufactured in a cleanroom to ensure cleanliness. They come with full documentation that confirms food grade and sterility.
Brand:  Sampling Systems™ 8630A-115S
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 17594676
  • £71.35 / Pack of 100
Estimated Shipment: 06-08-2024
to see stock.
Add to basket
Description
Description
  • Our disposable forceps are available in both sterile and non sterile versions.
  • They are suitable for lifting and moving small objects such as tablets, samples etc.
  • The handles have undulations moulded into them to give you extra grip.
  • These lightweight tweezers are suitable for food, chemical, pharmaceutical, or cosmetic applications.
  • These tweezers can be used straight from the bag.
  • The sterile tweezers have been processed by gamma irradiation.
  • Made from food grade PS.
  • Conforms to the latest FDA and EU requirements.
  • BSE/TSE free
  • To avoid contamination, they have been manufactured and then packed in a class 7 cleanroom.
  • They come with full documentation that confirms food grade and sterility.
Specifications
Specifications
Polystyrene
115 mm
Non-sterile
Bag