missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ SteriWare™ LiquiThief

Eliminate the risk of cross contamination when you use a disposable SteriWare™ LiquiThief. LiquiThief samplers are suitable for sampling a wide range of liquids and light oils.

Brand:  Sampling Systems™ 8050P-1000

Additional Details : Weight : 4.40000kg

 View more versions of this product

Product Code. 17504596

  • £155.00 / Pack of 20
Estimated Shipment: 12-09-2024
to see stock.
Add to basket

Description

Description

  • The LiquiThief is essentially a big syringe.
  • It is simple to use and comes with the added advantage that there is no cleaning validation and no cleaning.
  • Simply unwrap and use.
  • Pull the handle to take a sample.
  • The sampler is made in a cleanroom so you simply unwrap and use.
  • If you want to buy a disposable liquid sampler then the LiquiThief is the sampler of choice.
  • It is widely used to take samples of liquids, thin oils and ointments.
  • For more viscous materials use our SteriWare™ ViscoThief.
  • The disposable LiquiThief is a fast and convenient way of taking of liquid samples.
  • It is essentially a big syringe.
  • The operator pulls up the handle to pull the liquid into the sampler.
  • The further the handle is pulled the bigger the sample volume.
  • It is ideal for keg, container and drum sampling.
  • Ideal for quick sampling low viscosity liquids and oils.
  • All the polymers are food grade, FDA grade and EC compliant.
  • BSE/TSE free.
  • Full batch certificates are available.
  • Each sampler is manufactured and packed in a cleanroom to ensure that that they are clean until the point of use.
  • They are individually wrapped and can be used straight from the bag.
  • 3 lengths in both sterile and non sterile.
  • Available in both HDPE and PP construction.
Specifications

Specifications

Polypropylene
Yes
Individually Bagged
190 mL
Non-sterile
1000 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers