Learn More
Sampling Systems™ SteriWare™ Micro-Spatulas
SteriWare™ Micro-Spatulas are ideal for single use applications in pharmaceutical, chemical and other hygienic industries.
Brand: Sampling Systems™ 8015VB-170S
This item is not returnable.
View return policy
Description
- The disposable Micro Spatulas are ideal for a wide range of activities including sampling, dispensing and measuring small amounts of powder and granule.
- The narrow blades give you total control as well as enabling use in small spaces such as vials.
- The single-use spatula is ideal when costly cleaning and cleaning validation needs to be avoided.
- Simply unwrap the sampler and use it.
- It has been made in a controlled clean environment (class 100,000 cleanroom).
- They are available in white, blue and red.
- Colored versions are ideal if you are concerned about leaving it in the powder - they can be easily seen.
- Used by pharmaceutical, food and chemical companies around the world.
- Made from high-quality PS, PP or HDPE that conforms to the latest food grade, EC and FDA standards.
- Sterility Assurance Level (SAL) of 10-6
- BSE/TSE free.
- Batch certificates of conformity supplied.
Specifications
Polypropylene | |
Single-use | |
Blue |
0.3 mL | |
Sterile | |
Individually Bagged |