missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azlon™ Stirrer Bar Set
Resistant to virtually all chemicals
Brand: Azlon SWN515
241.05 GBP valid until 2024-07-30
Use promo code "21657" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.10000kg
Description
Resistant to virtually all chemicals
- Supplied in polarity aligned packs to ensure maximum coupling strength
- Assortment of the most popular types and sizes of PTFE magnetic stirrer bars
- Supplied in a red polypropylene compartmented box with opaque natural lid
Specifications
Pivot ring: two each of 12 x 4.5 25 x 6 45 x 8 and 60 x 9.5mm -Oval: two each of 20 x 10, 25 x 12, 35 x 16 and 50 x 20mm -Micro: three each of 7 x 2 and 10 x 3mm | |
Magnetic Stir Bar Set | |
PTFE |
PTFE | |
1/Ea. |