missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ Stirrer Motor with Electronic Controller
Use the geared shaft when you need higher torque and lower speeds and the direct-drive shaft when you need higher speeds and lower torque. Cole-Parmer™ Stirrer Motors with Electronic Controller offers smooth rotation and repeatability.
Brand: Cole-Parmer™ 04369-25
This item is not returnable.
View return policy
Description
- This stirrer motor features a totally enclosed 1⁄40hp motor and two output shafts
- Each shaft measures 5⁄16 in. dia.
- The versatile and reliable electronic motor controller features feedback circuitry that maintains speed even with viscosity changes
- Filtered circuitry ensures smooth rotation for repeatability in homogenizing and greatly reduces motor temperature and noise
- The reversing switch lets the motor shaft rotate in either direction and reverses pestle direction during homogenization
- The controller is connected to the motor with a cord so you can place it away from corrosive environments
Specifications
Homogenizer | |
4000 rpm | |
Develop a complete custom-fit tissue homogenizing system—choose a motor, pestle and tube | |
1 Ea. |
Special Offers
- Multiple Manufacturers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cole-Parmer™ Stirrer Motor with Electronic Controller > Speed: 4000rpm
Spot an opportunity for improvement?Share a Content Correction