Learn More
Scienceware Cross Magnetic Stir Bar Kit
Simplify your stirring with this Cross Magnetic Stir Bar Kit.
Brand: Scienceware 04765-00
Code : NEW
Additional Details : Weight : 0.27000kg
Description
- Chemically inert.
- Low friction.
- Leakproof PTFE coating.
- Kits each offer a wide variety of stir bar sizes to accomodate different glassware in the lab.
Specifications
5 Bars | |
Stirring Head Kit |