missing translation for 'onlineSavingsMsg'
Learn More

Stoddard Solvent

Product Code. 10474701 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
This item is not returnable. View return policy

Product Code. 10474701

Brand: Thermo Scientific Acros 419730010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 8052-41-3 | C8H15BrO2 | 223.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8052-41-3
Molecular Formula C8H15BrO2
Molecular Weight (g/mol) 223.11
MDL Number MFCD00132767
InChI Key IMSPZLCTPSXORJ-UHFFFAOYSA-N
Synonym 6-bromo-1-hexanol acetate, 1-acetoxy-6-bromohexane, stoddard solvent, 1-hexanol,6-bromo-,acetate, neo-clear r xylene substitute, w/w benzene, acetic acid 6-bromohexyl ester, 6-acetoxy-1-bromohexane
PubChem CID 11806291
IUPAC Name 6-bromohexyl acetate
SMILES CC(=O)OCCCCCCBr

Specifications

Melting Point -40.0°C
Density 0.7870g/mL
Boiling Point 149.0°C to 213.0°C
Quantity 1 kg
Flash Point >38°C
Packaging Glass Bottle
Physical Form Solvent
Specific Gravity 0.787
UN Number 1268
Solubility Information Solubility in water: insoluble.
Chemical Name or Material Stoddard solvent
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Stoddard solvent
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H372-Causes damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • H226-Flammable liquid and vapour.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P262-Do not get in eyes, on skin, or on clothing.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.