Learn More
Kinematica™ Stodger for Polymix™ MFC 90 D Mill
For active insertion of fibrous materials, which do not reach the grinding chamber due to their weight
Brand:  Kinematica™ 35080001
Product Code. 11940359
  • £42.80 / Each
Estimated Shipment: 03-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Stodger
Active insertion of fibrous materials, which do not reach the grinding chamber due to their weight
PX-MFC 90 D