Learn More
Seward™ Stomacher™ 80 Biomaster Blender
The Stomacher™ Model 80 Biomaster provide small tissue processing solutions for clinical, life science and microbiology applications.
Brand: Seward™ 0080/000/EU
Code : 03
Additional Details : Weight : 1.00000kg
Beschrijving
- Capable of processing samples from 250μL to 80mL, the Biomaster caters for the increased demand for smaller samples without reducing sample quality
- Allow samples to be processed safely and without risk of cross-contamination while maximizing visible cell recovery
- Adjustable process times and paddle speeds
- Proven enhancement in cell recovery and viability
- Cost-effective sample preparation
Specificatie
230 V | |
50 Hz | |
190 mm | |
For TB Work with Sputa and Tissues, Quality Control on Ointment and Cream Samples and Tissues in Animal Research |
230 V, 50 Hz | |
5 to 80 mL | |
290 mm |