Learn More
Stopcock Assembly and Cover for Nalgene™ Transparent Polycarbonate Classic Design Desiccator
GREENER_CHOICE
Get replacement parts for the Nalgene™ Transparent Polycarbonate Classic Design Desiccator
Brand:  Thermo Scientific Nalgene 715311-0250
286.73 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 10326231
  • £371.00 / Pack of 2
Estimated Shipment: 23-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Get the Stopcock Assembly and Cover replacement parts for the Nalgene™ Transparent Polycarbonate Classic Design Desiccator, Cat #5311-0250. The Nalgene™ Transparent Polycarbonate Classic Design Desiccator maintains an airtight seal without the need of vacuum grease and can be used under full-vacuum and non-vacuum conditions, and is an excellent alternative to glass.
Description:
  • Removable or replaceable stopcock
  • Port capped if removed (cap included)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Replacement Part
Cover for port on dessicator, polycarbonate