Learn More
Lenz Laborglasintrumente Stopcocks Straight Bore with PTFE Plug
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 02162910
 View more versions of this product
Product Code. 10222331
  • £117.00 / Each
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Acc. to DIN 12541-1
  • With PTFE plug and screwthread retaining device
  • One straight bore
  • Material: borosilicate glass 3.3
  • Leakage rate acc. to DIN 12540-1
Specifications
Specifications
10 mm
1-way stopcock
PTFE