Learn More
Kartell™ White Conical Stopper
Ensures sealing action on non-standard surfaces. Kartell™ White Conical Stopper features a tapered conical-shaped bottom to provide a leakproof seal.
Brand:  Kartell™ 0124700
Code : A0
Additional Details : Weight : 0.01000kg
Product Code. 10596191
  • £27.75 / Pack of 10
Estimated Shipment: 30-08-2024
to see stock.
Description
Description
  • Conical shaped
  • Seals non-standards surfaces
  • White in color
Specifications
Specifications
White
30.9mm
DIN 12242
NS 34/35
Polyethylene