missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ BS 2775 Series Rubber Stopper With One Hole
Shop All Fisherbrand Products
Click to view available options
Color:
Assorted
Unit Size:
Pack of 67
Description
- Small, tapered plug used to seal the openings of test tubes, flasks and other laboratory glassware
- Preferable for applications that require tighter seal or greater degree of chemical resistance
- Prevent contamination of samples by protecting contents of laboratory glassware from air
- Allows experimenters to shake or mix solutions without spilling them
Specifications
Specifications
| For Use With (Application) | For laboratory use |
| Quantity | 67/Pk. |
| Color | Assorted |
| Material | Rubber |
| Product Type | Tapered Stopper (2 holes) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction