Learn More
Wesbart™ Storage Bin
A useful way of keeping loose items safe within a freezer
Brand:  Wesbart™ BIN893
 View more versions of this product
Product Code. 15811458
  • £93.40 / Each
Estimated Shipment: 26-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Storage bins are manufactured from anodised aluminum or stainless steel
  • All storage bins have either a fold down handle or hand grip
  • Bins can be designed to have a handle on each end for transport if required
  • All storage bins are fitted with a label holder so the user can easily identify which bin they require
  • Specifications
    Specifications
    Storage Bin
    Freezer
    Handle on One End and a Label Holder
    232 mm
    Anodized Aluminum, Stainless Steel
    Keeping Loose Items Safe within a Freezer
    240 mm
    0.8 kg