missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Storage Box PF-945-ASB for Powered Air Respirator
3M™ Storage box PF-945-ASB for Powered Air Respirator System Asbestos Kit PF-602E-ASB.
Brand: 3M™ 7100265296
19.40 GBP valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Additional Details : Weight : 1.19600kg
Description
- 3M™ Storage box PF-945-ASB
- For use with Powered Air Respirator System Asbestos Kit PF-602E-ASB
Specifications
Storage Box | |
PF-945-ASB | |
Powered Air Respirator PF-600E-ASB | |
1 Ea. |