Learn More
Thermo Scientific™ Storerooms
Organize supplies, save valuable bench space using these six or nine drawer Storerooms. Order an empty cabinet or one fully stocked with vials and closures.
Brand:  Thermo Scientific™ C4000-MS
30.23 GBP valid until 2024-12-31
Use promo code "21693" to get your promotional price.
 View more versions of this product
Product Code. 12423578
  • £37.30 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Organize your supplies and save valuable bench space with Thermo Scientific™ National™ 8mm Standard Opening Screw Thread Vial Storerooms. Storerooms are available in six and nine drawer models. The six drawer mini-storeroom holds 500 vials and closures; the nine drawer full size storeroom holds 2000 vials and closures. Order an empty cabinet or one fully stocked with vials and closures.
  • Mini-Storeroom has 6 drawers; 5 drawers hold 1 vial pack each, 1 drawer holds 500 pre-stuffed DP caps
  • Full-size Storeroom has 9 deep drawers with a capacity of 2000 vials and pre-stuffed DP caps
  • Storerooms organize supplies and save bench space
  • Storeroom can be restocked as product is consumed
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Six-Drawer Mini Storeroom, Cabinet Only
1
6 Drawer Mini-Storeroom, Cabinet Only
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.