missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specac™ Storm™ 10 Short Pathlength Gas Cell FT-IR CaF2 Windows 47mm
Brand: Specac™
Code : Z3
Additional Details : Weight : 0.10000kg
Description
Pair of CaF2 Windows for use in a 10cm Storm Gas Cell GS05000 and GS05670.Specifications
10 cm Storm Gas Cell GS05000 and GS05670 |