missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ STP 120 Spin Tissue Processor
Process biological specimens from chemical fixation to paraffin infiltration with the Epredia™ STP 120 Spin Tissue Processor.
£24430.00 - £26310.00
Specifications
Basket Capacity | 120 or 240 Cassettes |
---|---|
Reagent Capacity | 1.8 L |
Diameter (English) | 33.5 in. |
Diameter (Metric) | 85 cm |
Height (English) | 19.6 to 27.5 in. |
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
11340710
|
Epredia™
813150 |
Includes ventilation system, 2 paraffin baths and 1 processing basket |
£26310.00
Each |
Estimated Shipment: 29-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12943705
|
Epredia™
813160 |
Includes ventilation system, 3 paraffin baths and 2 processing baskets |
£24430.00
Each |
Estimated Shipment: 09-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Immediate and delayed start processing modes
- Programmable spinning speed of either 60 or 70 rpm; spinning can also be programmed off
- Programmable immersion time in each station (from 1 minute to 99 hours: 59 minutes)
- Basket capacity of 120 or 240 cassettes
- Battery backup system in case of power failure
Performance:
Physical Specifications:
Includes:
Specifications
120 or 240 Cassettes | |
33.5 in. | |
19.6 to 27.5 in. | |
154 lb. | |
50/60 Hz |
1.8 L | |
85 cm | |
50 to 70 cm | |
70 kg | |
100/240 V |