Learn More
Masterflex™ Straight Barbed Fitting
These Masterflex™ Straight Barbed Fittings make tubing connections quick and easy, requiring no tools for installation.
Brand:  Masterflex™ 30612-13
Product Code. 12303800
  • £40.10 / Pack of 10
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Easily connect fluid paths without the use of tools
  • Slip flexible tubing or hose onto the barb to create a seal that typically holds pressures up to 30 psi (2 bar)
  • Available in a variety of sizes
Specifications
Specifications
HDPE
-53°C
Straight
0.25 in. Tubing I.D.
10 Pack
ETO sterilization