Learn More
Gosselin™ Straight Container, Hinged Cap
Product Code. 15482801
Click to view available options
Capacity (Metric):
35 mL
Unit Size:
Pack of 950
Product Code. 15482801
Brand: Gosselin™ FL3502
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Corning™ Gosselin™ Straight Container, Hinged Cap is lightweight and break-resistant.
  • Break-resistant polypropylene construction
  • Lightweight
  • Auto-stable hinged cap
  • Transparent, or blue
TRUSTED_SUSTAINABILITY
Specifications
Material Polypropylene
Capacity (Metric) 35 mL
Closure Type Hinged Cap
No. per Case 950/Cs.